4-Chloro-3-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

62260-500MG 62260-1G 62260-5G
77336-000EA 372.86 USD
77336-000 77355-816 77379-866
4-Chloro-3-methyl-1,1'-biphenyl ≥97%
4-Chloro-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁Cl
MW: 202.6794 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701661
CAS Number: 19482-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR