3-((2-Methoxybenzyl)oxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

62255-500MG 62255-1G 62255-5G 62255-25G
77331-562EA 225.57 USD
77331-562 77355-814 77367-702 77389-130
3-((2-Methoxybenzyl)oxy)benzaldehyde ≥97%
3-((2-Methoxybenzyl)oxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.2698 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11891658
CAS Number: 123926-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR