4-Fluoro-2-((2-methoxybenzyl)oxy)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62254-500MG 62254-1G 62254-5G 62254-25G
77331-560EA 152.44 USD
77331-560 77345-478 77367-700 77389-128
4-Fluoro-2-((2-methoxybenzyl)oxy)-1-methylbenzene ≥97%
4-Fluoro-2-((2-methoxybenzyl)oxy)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₂
Storage Temperature: Ambient
MDL Number: MFCD34760772
CAS Number: 1975638-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR