3-Bromo-6-chloro-[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

62232-500MG 62232-1G 62232-5G
77337-036EA 406.85 USD
77337-036 77355-812 77379-864
3-Bromo-6-chloro-[1,2,4]triazolo[4,3-a]pyridine ≥97%
3-Bromo-6-chloro-[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrClN₃
MW: 232.4651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28368650
CAS Number: 1427852-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR