1,3-Dibromo-2-chloro-4-fluoro-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

62231-500MG 62231-1G 62231-5G 62231-25G
77331-558EA 271.92 USD
77331-558 77355-810 77367-698 77389-126
1,3-Dibromo-2-chloro-4-fluoro-5-methylbenzene ≥95%
1,3-Dibromo-2-chloro-4-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₂ClF
Storage Temperature: Ambient
MDL Number: MFCD34760769
CAS Number: 2901083-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR