4'-Methoxy-5-methyl-[1,1'-biphenyl]-2-amine ≥97%

Supplier: AOBChem USA

Warning

62202-500MG 62202-1G 62202-5G
77331-554EA 307.97 USD
77331-554 77355-806 77379-858
4'-Methoxy-5-methyl-[1,1'-biphenyl]-2-amine ≥97%
4'-Methoxy-5-methyl-[1,1'-biphenyl]-2-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO
MW: 213.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11933369
CAS Number: 637345-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR