4',5-Dimethyl-[1,1'-biphenyl]-2-amine ≥97%

Supplier: AOBChem USA

Warning

62200-500MG 62200-1G 62200-5G 62200-25G
77331-552EA 123.6 USD
77331-552 77344-036 77367-694 77379-856
4',5-Dimethyl-[1,1'-biphenyl]-2-amine ≥97%
4',5-Dimethyl-[1,1'-biphenyl]-2-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD12824037
CAS Number: 116668-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR