4-(5-Methyl-2-nitrophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

62198-500MG 62198-1G 62198-5G
77331-548EA 307.97 USD
77331-548 77355-802 77379-852
4-(5-Methyl-2-nitrophenyl)pyridine ≥97%
4-(5-Methyl-2-nitrophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34760764
CAS Number: 2764731-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR