Methyl 2-(1-hydroxycycloheptyl)acetate ≥97%

Supplier: AOBChem USA

Warning

62184-500MG 62184-1G 62184-5G
77337-762EA 440.84 USD
77337-762 77355-794 77379-846
Methyl 2-(1-hydroxycycloheptyl)acetate ≥97%
Methyl 2-(1-hydroxycycloheptyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD30604731
CAS Number: 61704-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR