Ethyl 2,3-dibromo-5-fluoro-4-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

62174-500MG 62174-1G 62174-5G 62174-25G
77331-542EA 128.75 USD
77331-542 77344-278 77367-688 77379-842
Ethyl 2,3-dibromo-5-fluoro-4-methylbenzoate ≥95%
Ethyl 2,3-dibromo-5-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉Br₂FO₂
MW: 339.9836 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760752
CAS Number: 2764728-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR