Methyl 2,3-dibromo-5-fluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

62173-500MG 62173-1G 62173-5G 62173-25G
77331-540EA 112.27 USD
77331-540 77343-334 77367-686 77379-840
Methyl 2,3-dibromo-5-fluoro-4-methylbenzoate ≥97%
Methyl 2,3-dibromo-5-fluoro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇Br₂FO₂
MW: 325.9570 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34647007
CAS Number: 2324177-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR