2,3-Dibromo-5-fluoro-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

62172-500MG 62172-1G 62172-5G 62172-25G
77331-538EA 108.15 USD
77331-538 77343-186 77367-684 77379-838
2,3-Dibromo-5-fluoro-4-methylbenzoic acid ≥95%
2,3-Dibromo-5-fluoro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅Br₂FO₂
MW: 311.9305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760751
CAS Number: 2764733-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR