1-Butoxy-3-iodo-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

62162-500MG 62162-1G 62162-5G
77336-844EA 394.49 USD
77336-844 77355-788 77379-832
1-Butoxy-3-iodo-5-(trifluoromethyl)benzene ≥97%
1-Butoxy-3-iodo-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₃IO
MW: 344.1120 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760747
CAS Number: 2764731-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR