(3-Butoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

62161-500MG 62161-1G 62161-5G
77331-534EA 326.51 USD
77331-534 77355-786 77379-830
(3-Butoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(3-Butoxy-5-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₂H₁₅F₃OS
Storage Temperature: Ambient
MDL Number: MFCD34760746
CAS Number: 2807471-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR