3-Amino-5-bromo-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62151-500MG 62151-1G 62151-5G 62151-25G
77331-532EA 163.77 USD
77331-532 77346-096 77367-678 77389-120
3-Amino-5-bromo-4-methylbenzoic acid ≥97%
3-Amino-5-bromo-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24114619
CAS Number: 34545-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR