1-Bromo-3-ethoxy-2-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62131-500MG 62131-1G 62131-5G 62131-25G
77331-518EA 161.71 USD
77331-518 77345-908 77367-664 77389-108
1-Bromo-3-ethoxy-2-methoxy-5-methylbenzene ≥97%
1-Bromo-3-ethoxy-2-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MW: 245.1130 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760729
CAS Number: 2386135-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR