1-(2-Bromo-5-iodophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

62124-500MG 62124-1G 62124-5G
77337-690EA 436.72 USD
77337-690 77355-772 77379-820
1-(2-Bromo-5-iodophenyl)ethanol ≥97%
1-(2-Bromo-5-iodophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈BrIO
Storage Temperature: Ambient
MDL Number: MFCD32647880
CAS Number: 1691038-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR