1-(5-Chloro-2-fluoro-3-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

62112-500MG 62112-1G 62112-5G 62112-25G
77331-510EA 187.46 USD
77331-510 77355-764 77367-658 77389-102
1-(5-Chloro-2-fluoro-3-methoxyphenyl)ethanone ≥97%
1-(5-Chloro-2-fluoro-3-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.6100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28526635
CAS Number: 1785603-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR