5,7-Dibromo-2-methyl-2H-indazole ≥97%

Supplier: AOBChem USA

Warning

62108-500MG 62108-1G 62108-5G
77337-192EA 408.91 USD
77337-192 77355-762 77379-810
5,7-Dibromo-2-methyl-2H-indazole ≥97%
5,7-Dibromo-2-methyl-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₆Br₂N₂
MW: 289.9546 g/mol
Storage Temperature: Ambient
CAS Number: 1644115-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR