5,7-Dibromo-1-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

62107-500MG 62107-1G 62107-5G
77339-098EA 550.02 USD
77339-098 77355-760 77379-808
5,7-Dibromo-1-methyl-1H-indazole ≥97%
5,7-Dibromo-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₆Br₂N₂
MW: 289.9546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29089649
CAS Number: 1644115-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR