2-Bromo-4-chloro-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

62078-500MG 62078-1G 62078-5G
77338-642EA 505.73 USD
77338-642 77355-758 77379-806
2-Bromo-4-chloro-5-methylbenzoic acid ≥95%
2-Bromo-4-chloro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.4890 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760712
CAS Number: 2090689-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR