1,2,3-Tribromo-4-chloro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62068-500MG 62068-1G 62068-5G 62068-25G
77331-508EA 99.91 USD
77331-508 77342-744 77367-654 77379-800
1,2,3-Tribromo-4-chloro-5-methylbenzene ≥97%
1,2,3-Tribromo-4-chloro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₃Cl
Storage Temperature: Ambient
MDL Number: MFCD34760707
CAS Number: 2764730-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR