1,3-Dibromo-4-chloro-2-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62066-500MG 62066-1G 62066-5G 62066-25G
77331-506EA 120.51 USD
77331-506 77343-776 77367-652 77379-798
1,3-Dibromo-4-chloro-2-iodo-5-methylbenzene ≥97%
1,3-Dibromo-4-chloro-2-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Br₂ClI
Storage Temperature: Ambient
MDL Number: MFCD00052445
CAS Number: 1000572-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR