3,5-Dibromo-2-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62064-1G 62064-5G 62064-25G
77341-696EA 100.94 USD
77341-696 77367-648 77379-794
3,5-Dibromo-2-chlorobenzoic acid ≥97%
3,5-Dibromo-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Br₂ClO₂
Storage Temperature: Ambient
MDL Number: MFCD07780692
CAS Number: 27003-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR