4,5-Dibromo-2-chlorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
62044-500MG
62044-1G
62044-5G
77338-052EA
460.41
USD
77338-052
77355-752
77379-790
4,5-Dibromo-2-chlorobenzoic acid ≥97%
4,5-Dibromo-2-chlorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃Br₂ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD24107219 CAS Number: 1509491-97-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...