2-Bromo-3,6-dichloropyridine ≥95%

Supplier: AOBChem USA

Warning

62023-500MG 62023-1G 62023-25G 62023-5G
77331-498EA 301.79 USD
77331-498 77355-748 77389-098 77379-786
2-Bromo-3,6-dichloropyridine ≥95%
2-Bromo-3,6-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
MW: 226.8860 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447566
CAS Number: 1266119-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR