2,6-Dibromo-3-fluoro-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

62014-500MG 62014-1G 62014-5G 62014-25G
77331-496EA 191.58 USD
77331-496 77355-746 77367-644 77389-096
2,6-Dibromo-3-fluoro-4-(methylthio)pyridine ≥97%
2,6-Dibromo-3-fluoro-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₄Br₂FNS
MW: 300.9740 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760686
CAS Number: 2764728-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR