2,6-Dibromo-3-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

62013-500MG 62013-1G 62013-5G 62013-25G
77331-494EA 181.28 USD
77331-494 77346-868 77367-642 77389-094
2,6-Dibromo-3-fluoro-4-iodopyridine ≥97%
2,6-Dibromo-3-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅HBr₂FIN
Storage Temperature: Ambient
MDL Number: MFCD34760685
CAS Number: 2764731-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR