2',3'-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

61977-500MG 61977-1G 61977-5G
77338-350EA 479.98 USD
77338-350 77355-734 77379-780
2',3'-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2',3'-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O
MW: 232.2254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32110992
CAS Number: 1926893-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR