2,3,4'-Trifluoro-4-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61976-500MG 61976-1G 61976-5G
77337-026EA 405.82 USD
77337-026 77355-732 77379-778
2,3,4'-Trifluoro-4-methyl-1,1'-biphenyl ≥97%
2,3,4'-Trifluoro-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉F₃
MW: 222.2057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760662
CAS Number: 2194613-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR