2,3-Difluoro-4-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61973-500MG 61973-1G 61973-5G
77337-736EA 439.81 USD
77337-736 77355-728 77379-774
2,3-Difluoro-4-methyl-1,1'-biphenyl ≥97%
2,3-Difluoro-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂
MW: 204.2153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23102312
CAS Number: 1824585-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR