4-Chloro-2-fluoro-3-iodophenol ≥97%

Supplier: AOBChem USA

Warning

61968-500MG 61968-1G 61968-5G
77335-708EA 363.59 USD
77335-708 77355-724 77379-770
4-Chloro-2-fluoro-3-iodophenol ≥97%
4-Chloro-2-fluoro-3-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃ClFIO
MW: 272.4432 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760655
CAS Number: 2386540-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR