tert-Butyl 4-(4-iodopyridin-2-yl)piperazine-1-carboxylate ≥97%

Supplier: AOBChem USA

Warning

61965-500MG 61965-1G 61965-5G 61965-25G
77331-480EA 236.9 USD
77331-480 77355-722 77367-632 77389-084
tert-Butyl 4-(4-iodopyridin-2-yl)piperazine-1-carboxylate ≥97%
tert-Butyl 4-(4-iodopyridin-2-yl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀IN₃O₂
MW: 389.2319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28400243
CAS Number: 1704065-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR