1-(3-(Benzyloxy)-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

61957-500MG 61957-1G 61957-25G 61957-5G
77331-478EA 299.73 USD
77331-478 77355-720 77389-082 77379-768
1-(3-(Benzyloxy)-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-(Benzyloxy)-6-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₅H₉ClF₄O₂
MW: 332.6773 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760649
CAS Number: 2807467-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR