3-(Benzyloxy)-6-chloro-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61950-500MG 61950-1G 61950-5G
77331-476EA 347.11 USD
77331-476 77355-718 77379-766
3-(Benzyloxy)-6-chloro-2-fluorobenzoic acid ≥97%
3-(Benzyloxy)-6-chloro-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34760642
CAS Number: 1034026-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR