1-Chloro-4-ethoxy-3-fluoro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

61947-500MG 61947-1G 61947-5G
77337-788EA 443.93 USD
77337-788 77355-714 77379-762
1-Chloro-4-ethoxy-3-fluoro-2-methylbenzene ≥95%
1-Chloro-4-ethoxy-3-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO
MW: 188.6265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751535
CAS Number: 1807206-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR