2-Ethoxy-3-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61917-500MG 61917-1G 61917-5G
77340-142EA 791.04 USD
77340-142 77355-700 77379-750
2-Ethoxy-3-fluoro-5-methylbenzoic acid ≥97%
2-Ethoxy-3-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.1909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28626778
CAS Number: 1555623-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR