2-(Benzyloxy)-1-bromo-3-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61913-500MG 61913-1G 61913-5G 61913-25G
77331-464EA 112.27 USD
77331-464 77343-310 77367-626 77379-746
2-(Benzyloxy)-1-bromo-3-fluoro-5-methylbenzene ≥97%
2-(Benzyloxy)-1-bromo-3-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.1468 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760623
CAS Number: 2807455-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR