4-Bromo-3-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61907-500MG 61907-1G 61907-5G
77331-460EA 307.97 USD
77331-460 77355-696 77379-744
4-Bromo-3-methoxy-4'-methyl-1,1'-biphenyl ≥97%
4-Bromo-3-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
MW: 277.1564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760619
CAS Number: 2764733-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR