Methyl 4-bromo-3-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

61892-500MG 61892-1G 61892-5G 61892-25G
77331-450EA 119.48 USD
77331-450 77343-616 77367-614 77379-738
Methyl 4-bromo-3-isopropylbenzoate ≥97%
Methyl 4-bromo-3-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30737874
CAS Number: 2169639-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR