4-(4-Bromo-3,5-difluorophenyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

61868-500MG 61868-1G 61868-5G 61868-25G
77331-448EA 278.1 USD
77331-448 77355-690 77367-612 77389-068
4-(4-Bromo-3,5-difluorophenyl)morpholine ≥97%
4-(4-Bromo-3,5-difluorophenyl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrF₂NO
MW: 278.0933 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760612
CAS Number: 2028864-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR