4-(3,5-Difluorophenyl)morpholine ≥97%
Supplier: AOBChem USA
|
Warning
|
61867-500MG
61867-1G
61867-5G
61867-25G
77331-446EA
254.41
USD
77331-446
77355-688
77367-610
77389-066
4-(3,5-Difluorophenyl)morpholine ≥97%
4-(3,5-Difluorophenyl)morpholine
Precaución: for Research Use Only
Formula:
C₁₀H₁₁F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD27928097 CAS Number: 736991-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...