1-(3,5-Difluorophenyl)pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

61866-500MG 61866-1G 61866-5G 61866-25G
77331-444EA 164.8 USD
77331-444 77346-148 77367-608 77389-064
1-(3,5-Difluorophenyl)pyrrolidine ≥97%
1-(3,5-Difluorophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂N
MW: 183.1978 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31768710
CAS Number: 1314137-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR