4-(3,5-Difluorophenyl)thiomorpholine ≥97%

Supplier: AOBChem USA

Warning

61864-500MG 61864-1G 61864-5G 61864-25G
77331-442EA 281.19 USD
77331-442 77355-686 77367-606 77389-062
4-(3,5-Difluorophenyl)thiomorpholine ≥97%
4-(3,5-Difluorophenyl)thiomorpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₂NS
MW: 215.2628 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760611
CAS Number: 2807470-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR