2-Methoxy-N-methyl-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61860-500MG 61860-1G 61860-5G 61860-25G
77331-438EA 103 USD
77331-438 77343-044 77367-602 77379-736
2-Methoxy-N-methyl-5-(trifluoromethyl)benzamide ≥97%
2-Methoxy-N-methyl-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34760610
CAS Number: 2764729-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR