2-Methoxy-N,N-dimethyl-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61859-500MG 61859-1G 61859-5G 61859-25G
77331-436EA 103 USD
77331-436 77343-042 77367-600 77379-734
2-Methoxy-N,N-dimethyl-5-(trifluoromethyl)benzamide ≥97%
2-Methoxy-N,N-dimethyl-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD29097427
CAS Number: 1369783-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR