N-Cyclopropyl-2-methoxy-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61858-500MG 61858-1G 61858-5G 61858-25G
77331-434EA 152.44 USD
77331-434 77345-476 77367-598 77389-058
N-Cyclopropyl-2-methoxy-5-(trifluoromethyl)benzamide ≥97%
N-Cyclopropyl-2-methoxy-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34760609
CAS Number: 2764729-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR