N-Cyclohexyl-2-methoxy-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

61857-500MG 61857-1G 61857-5G 61857-25G
77331-432EA 144.2 USD
77331-432 77345-022 77367-596 77389-056
N-Cyclohexyl-2-methoxy-5-(trifluoromethyl)benzamide ≥97%
N-Cyclohexyl-2-methoxy-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₅H₁₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34760608
CAS Number: 2764731-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR