(2-Methoxy-5-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

61856-500MG 61856-1G 61856-5G 61856-25G
77331-430EA 99.91 USD
77331-430 77342-742 77367-594 77379-732
(2-Methoxy-5-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥97%
(2-Methoxy-5-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₄F₃NO₂
MW: 273.2509 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760607
CAS Number: 2764730-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR