2-Chloro-4-(p-tolyl)nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

61850-500MG 61850-1G 61850-5G
77336-438EA 387.28 USD
77336-438 77355-682 77379-728
2-Chloro-4-(p-tolyl)nicotinaldehyde ≥97%
2-Chloro-4-(p-tolyl)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClNO
MW: 231.6776 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760602
CAS Number: 2764728-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR